4-Bromo-1-iodo-2-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-iodonitrobenzene

HC-2846-5G HC-2846-1G
COBBHC-2846-5GEA 0 GBP
COBBHC-2846-5G COBBHC-2846-1G
4-Bromo-1-iodo-2-nitrobenzene
4-Bromo-1-iodo-2-nitrobenzene
Formula: C₆H₃BrINO₂
MW: 327.9020 g/mol
MDL Number: MFCD11505492
CAS Number: 112671-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR