5-Bromo-2-(tert-butylamino)-4-fluoro-1-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1454-5G AN-1454-25G AN-1454-1G
COBBAN-1454-5GEA 0 GBP
COBBAN-1454-5G COBBAN-1454-25G COBBAN-1454-1G
5-Bromo-2-(tert-butylamino)-4-fluoro-1-nitrobenzene
5-Bromo-2-(tert-butylamino)-4-fluoro-1-nitrobenzene
Formula: C₁₀H₁₂BrFN₂O₂
MDL Number: MFCD12025955
CAS Number: 1150114-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR