2-Bromo-5-fluoro-N-phenylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4436-5G CA-4436-1G
COBBCA-4436-5GEA 0 GBP
COBBCA-4436-5G COBBCA-4436-1G
2-Bromo-5-fluoro-N-phenylbenzamide
2-Bromo-5-fluoro-N-phenylbenzamide
Formula: C₁₃H₉BrFNO
MDL Number: MFCD09800907
CAS Number: 949443-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR