2-Bromo-N-tert-butyl-5-fluorobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4394-1G CA-4394-5G
COBBCA-4394-1GEA 0 GBP
COBBCA-4394-1G COBBCA-4394-5G
2-Bromo-N-tert-butyl-5-fluorobenzamide
2-Bromo-N-tert-butyl-5-fluorobenzamide
Formula: C₁₁H₁₃BrFNO
MDL Number: MFCD09475888
CAS Number: 941294-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR