3-Bromo-4-(trifluoromethoxy)benzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4780-5G CA-4780-1G
COBBCA-4780-5GEA 0 GBP
COBBCA-4780-5G COBBCA-4780-1G
3-Bromo-4-(trifluoromethoxy)benzamide
3-Bromo-4-(trifluoromethoxy)benzamide
Formula: C₈H₅BrF₃NO₂
MDL Number: MFCD08458067
CAS Number: 914636-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR