4-Bromo-N-ethyl-3-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4652-5G CA-4652-1G
COBBCA-4652-5GEA 0 GBP
COBBCA-4652-5G COBBCA-4652-1G
4-Bromo-N-ethyl-3-methoxybenzamide
4-Bromo-N-ethyl-3-methoxybenzamide
Formula: C₁₀H₁₂BrNO₂
MDL Number: MFCD11504876
CAS Number: 1072944-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR