4-Bromo-N-isopropyl-3-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4653-5G CA-4653-1G
COBBCA-4653-5GEA 0 GBP
COBBCA-4653-5G COBBCA-4653-1G
4-Bromo-N-isopropyl-3-methoxybenzamide
4-Bromo-N-isopropyl-3-methoxybenzamide
Formula: C₁₁H₁₄BrNO₂
MDL Number: MFCD11504877
CAS Number: 1072944-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR