2-Bromo-N,N-dimethylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4728-5G
CA-4728-1G
CA-4728-25G
COBBCA-4728-5GEA
0
GBP
COBBCA-4728-5G
COBBCA-4728-1G
COBBCA-4728-25G
2-Bromo-N,N-dimethylbenzamide
2-Bromo-N,N-dimethylbenzamide
Formula:
C₉H₁₀BrNO MW: 228.0900 g/mol |
MDL Number:
MFCD00463635 CAS Number: 54616-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...