4-Bromo-N,N-dimethylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4418-5G CA-4418-25G CA-4418-1G
COBBCA-4418-5GEA 0 GBP
COBBCA-4418-5G COBBCA-4418-25G COBBCA-4418-1G
4-Bromo-N,N-dimethylbenzamide
4-Bromo-N,N-dimethylbenzamide
Formula: C₉H₁₀BrNO
MW: 228.0900 g/mol
MDL Number: MFCD00463696
CAS Number: 18469-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR