2-Bromo-N,N-diisopropylbenzamide

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBCS-9416-1GEA 0 GBP
COBBCS-9416-1G COBBCS-9416-5G
2-Bromo-N,N-diisopropylbenzamide
2-Bromo-N,N-diisopropylbenzamide
Formula: C₁₃H₁₈BrNO
MDL Number: MFCD00583268
CAS Number: 79839-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR