5-Bromo-2-chloro-N-phenylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4704-1G CA-4704-25G CA-4704-5G
COBBCA-4704-1GEA 0 GBP
COBBCA-4704-1G COBBCA-4704-25G COBBCA-4704-5G
5-Bromo-2-chloro-N-phenylbenzamide
5-Bromo-2-chloro-N-phenylbenzamide
Formula: C₁₃H₉BrClNO
MDL Number: MFCD02210174
CAS Number: 420826-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR