5-Bromo-2-chloro-N-cyclopropylbenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4723-5G CA-4723-1G
COBBCA-4723-5GEA 0 GBP
COBBCA-4723-5G COBBCA-4723-1G
5-Bromo-2-chloro-N-cyclopropylbenzamide
5-Bromo-2-chloro-N-cyclopropylbenzamide
Formula: C₁₀H₉BrClNO
MW: 274.5416 g/mol
MDL Number: MFCD04226747
CAS Number: 669734-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR