3-(tert-Butylamino)propionitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(tert-Butylamino)propanenitrile

018333-10G 018333-25G
FLUO018333-10GEA 0 GBP
FLUO018333-10G FLUO018333-25G
3-(tert-Butylamino)propionitrile
3-(tert-Butylamino)propionitrile
Formula: C₇H₁₄N₂
MW: 126.1995 g/mol
MDL Number: MFCD00045637
CAS Number: 21539-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR