2-(4-Nitrophenyl)oxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3339-5G HC-3339-1G
COBBHC-3339-5GEA 0 GBP
COBBHC-3339-5G COBBHC-3339-1G
2-(4-Nitrophenyl)oxazole
2-(4-Nitrophenyl)oxazole
Formula: C₉H₆N₂O₃
MDL Number: MFCD08276758
CAS Number: 62882-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR