Ethyl-5-(2,4-dichlorophenyl)-4-oxazolecarboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6228-5G
HC-6228-1G
COBBHC-6228-5GEA
0
GBP
COBBHC-6228-5G
COBBHC-6228-1G
Ethyl-5-(2,4-dichlorophenyl)-4-oxazolecarboxylate
Ethyl 5-(2,4-dichlorophenyl)-4-oxazolecarboxylate
Formula:
C₁₂H₉Cl₂NO₃ |
MDL Number:
MFCD00105347 CAS Number: 254749-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...