Ethyl-5-(2,3-dichlorophenyl)-4-oxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6229-5G HC-6229-1G
COBBHC-6229-5GEA 0 GBP
COBBHC-6229-5G COBBHC-6229-1G
Ethyl-5-(2,3-dichlorophenyl)-4-oxazolecarboxylate
Ethyl 5-(2,3-dichlorophenyl)-4-oxazolecarboxylate
Formula: C₁₂H₉Cl₂NO₃
MDL Number: MFCD09800991
CAS Number: 951885-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR