5-(4-Formylphenyl)-1H-tetrazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(1H-Tetrazol-5-yl)benzaldehyde

AS23655G5 AS23655G1
ACTIAS23655G5EA 462 GBP
ACTIAS23655G5 ACTIAS23655G1
5-(4-Formylphenyl)-1H-tetrazole
5-(4-Formylphenyl)-1H-tetrazole
Formula: C₈H₆N₄O
MW: 174.16 g/mol
MDL Number: MFCD01311033
CAS Number: 74815-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR