5-(Ethylthio)-1H-tetrazole (ETT)
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Ethyltio-1H-Tetrazole
MOLE51057635-25GEA
0
GBP
MOLE51057635-25G
5-(Ethylthio)-1H-tetrazole (ETT)
5-(Ethylthio)-1H-tetrazole (ETT)
Formula:
C₃H₆N₄S MW: 130.17 g/mol |
MDL Number:
MFCD00068733 CAS Number: 89797-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...