Ethyl-2-chloro-4-oxazolecarboxylate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 2-chlorooxazole-4-carboxylate
HI-1195-5G
HI-1195-1G
COBBHI-1195-5GEA
0
GBP
COBBHI-1195-5G
COBBHI-1195-1G
Ethyl-2-chloro-4-oxazolecarboxylate
Ethyl 2-chloro-4-oxazolecarboxylate
Formula:
C₆H₆ClNO₃ MW: 175.57 g/mol |
MDL Number:
MFCD06660120 CAS Number: 460081-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...