5-(4-Fluorophenyl)isoxazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6883-5G HC-6883-1G HC-6883-25G
COBBHC-6883-5GEA 0 GBP
COBBHC-6883-5G COBBHC-6883-1G COBBHC-6883-25G
5-(4-Fluorophenyl)isoxazole
5-(4-Fluorophenyl)isoxazole
Formula: C₉H₆FNO
MW: 163.15 g/mol
MDL Number: MFCD02183544
CAS Number: 138716-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR