3-(4-Chlorophenyl)-5-isoxazolecarboxylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

11L-366S.500 11L-366S.1 11L-366S.5 11L-366S.10
BIOT11L-366S.500EA 0 GBP
BIOT11L-366S.500 BIOT11L-366S.1 BIOT11L-366S.5 BIOT11L-366S.10
3-(4-Chlorophenyl)-5-isoxazolecarboxylic acid
3-(4-Chlorophenyl)-5-isoxazolecarboxylic acid
Formula: C₁₀H₆ClNO₃
MDL Number: MFCD01935955
CAS Number: 338982-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR