Methyl-5-(3-chlorophenyl)-4-isoxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2040-5G HC-2040-1G
COBBHC-2040-5GEA 0 GBP
COBBHC-2040-5G COBBHC-2040-1G
Methyl-5-(3-chlorophenyl)-4-isoxazolecarboxylate
Methyl 5-(3-chlorophenyl)-4-isoxazolecarboxylate
Formula: C₁₁H₈ClNO₃
MDL Number: MFCD11504883
CAS Number: 1065074-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR