Methyl-4-(4-chlorophenyl)-5-isoxazolecarboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1438-5G HI-1438-1G
COBBHI-1438-5GEA 0 GBP
COBBHI-1438-5G COBBHI-1438-1G
Methyl-4-(4-chlorophenyl)-5-isoxazolecarboxylate
Methyl 4-(4-chlorophenyl)-5-isoxazolecarboxylate
Formula: C₁₁H₈ClNO₃
MDL Number: MFCD11504938
CAS Number: 1072944-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR