5-Methoxy-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2665-5G HC-2665-1G
COBBHC-2665-5GEA 0 GBP
COBBHC-2665-5G COBBHC-2665-1G
5-Methoxy-1H-indazole
5-Methoxy-1H-indazole
Formula: C₈H₈N₂O
MW: 148.1600 g/mol
MDL Number: MFCD07781657
CAS Number: 94444-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR