3-Iodo-5-nitro-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1616-1G HI-1616-5G
COBBHI-1616-1GEA 0 GBP
COBBHI-1616-1G COBBHI-1616-5G
3-Iodo-5-nitro-1H-indazole
3-Iodo-5-nitro-1H-indazole
Formula: C₇H₄IN₃O₂
MW: 289.0300 g/mol
MDL Number: MFCD07781630
CAS Number: 70315-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR