6-Amino-3-iodo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Iodo-1H-indazol-6-amine

AS12490G1 AS12490G5
ACTIAS12490G1EA 203 GBP
ACTIAS12490G1 ACTIAS12490G5
6-Amino-3-iodo-1H-indazole
6-Amino-3-iodo-1H-indazole
Formula: C₇H₆IN₃
MW: 259.471 g/mol
MDL Number: MFCD07781651
CAS Number: 885519-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR