3-Iodo-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Iodoindazole

HI-1615-5G HI-1615-1G
COBBHI-1615-5GEA 0 GBP
COBBHI-1615-5G COBBHI-1615-1G
3-Iodo-1H-indazole
3-Iodo-1H-indazole
Formula: C₇H₅IN₂
MW: 244.0325 g/mol
MDL Number: MFCD04972469
CAS Number: 66607-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR