5-Amino-7-bromo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-Bromo-1H-indazol-5-amine

ACTIAS5170G1EA 0 GBP
ACTIAS5170G1 ACTIAS5170G5
5-Amino-7-bromo-1H-indazole
5-Amino-7-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
MDL Number: MFCD08275114
CAS Number: 953411-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR