Methyl-3-bromo-1H-indazole-5-carboxylate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 3-bromo-1H-indazole-5-carboxylate
AS12119G1
AS12119G5
ACTIAS12119G1EA
30
GBP
ACTIAS12119G1
ACTIAS12119G5
Methyl-3-bromo-1H-indazole-5-carboxylate
Methyl 3-bromo-1H-indazole-5-carboxylate
Formula:
C₉H₇BrN₂O₂ MW: 255.0681 g/mol |
MDL Number:
MFCD11045401 CAS Number: 1086391-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...