1H-Indazole-7-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

AS12274G1 AS12274G5
ACTIAS12274G1EA 113 GBP
ACTIAS12274G1 ACTIAS12274G5
1H-Indazole-7-boronic acid pinacol ester
1H-Indazole-7-boronic acid pinacol ester
Formula: C₁₃H₁₇BN₂O₂
MW: 244.0973 g/mol
MDL Number: MFCD08059264
CAS Number: 915411-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR