Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

008297-5G 008297-1G
FLUO008297-5GEA 0 GBP
FLUO008297-5G FLUO008297-1G
Ethyl-5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₂FN₃O₂
MW: 249.2410 g/mol
MDL Number: MFCD00173917
CAS Number: 138907-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR