5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

017437-5G 017437-10G 017437-25G 017437-1G
FLUO017437-5GEA 0 GBP
FLUO017437-5G FLUO017437-10G FLUO017437-25G FLUO017437-1G
5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole
5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole
Formula: C₅H₆ClN₃O₂
MW: 175.58 g/mol
MDL Number: MFCD00052534
CAS Number: 13551-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR