5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12F-330S.500
12F-330S.1
12F-330S.5
12F-330S.10
BIOT12F-330S.500EA
0
GBP
BIOT12F-330S.500
BIOT12F-330S.1
BIOT12F-330S.5
BIOT12F-330S.10
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid
Formula:
C₆H₇ClN₂O₂ |
MDL Number:
MFCD00232043 CAS Number: 27006-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...