6-Cyano-1-ethylbenzimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1597-1G HI-1597-5G
COBBHI-1597-1GEA 0 GBP
COBBHI-1597-1G COBBHI-1597-5G
6-Cyano-1-ethylbenzimidazole
6-Cyano-1-ethylbenzimidazole
Formula: C₁₀H₉N₃
MDL Number: MFCD13195705
CAS Number: 1215205-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR