5-Bromo-2-methylbenzimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-methyl-1H-benzimidazole, 5-Bromo-2-methyl-1H-benzo[d]imidazole

HI-1581-5G HI-1581-1G
COBBHI-1581-5GEA 0 GBP
COBBHI-1581-5G COBBHI-1581-1G
5-Bromo-2-methylbenzimidazole
5-Bromo-2-methylbenzimidazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD00457521
CAS Number: 1964-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR