3-Chloro-6-(4-iodo-1H-pyrazol-1-yl)pyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6100-5G HC-6100-1G
COBBHC-6100-5GEA 0 GBP
COBBHC-6100-5G COBBHC-6100-1G
3-Chloro-6-(4-iodo-1H-pyrazol-1-yl)pyridazine
3-Chloro-6-(4-iodo-1H-pyrazol-1-yl)pyridazine
Formula: C₇H₄ClIN₄
MDL Number: MFCD09475885
CAS Number: 957035-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR