3-Iodo-4-methyl-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1614-5G
HI-1614-1G
COBBHI-1614-5GEA
0
GBP
COBBHI-1614-5G
COBBHI-1614-1G
3-Iodo-4-methyl-1H-indazole
3-Iodo-4-methyl-1H-indazole
Formula:
C₈H₇IN₂ MW: 258.0591 g/mol |
MDL Number:
MFCD07781537 CAS Number: 885522-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...