7-Fluoro-1-methyl-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0202-5G IN-0202-1G IN-0202-250M
COBBIN-0202-5GEA 0 GBP
COBBIN-0202-5G COBBIN-0202-1G COBBIN-0202-250M
7-Fluoro-1-methyl-1H-indazole
7-Fluoro-1-methyl-1H-indazole
Formula: C₈H₇FN₂
MW: 150.1500 g/mol
MDL Number: MFCD12546530
CAS Number: 1187386-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR