1-Acetyl-4-bromo-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21871G1 AS21871G5
ACTIAS21871G1EA 0 GBP
ACTIAS21871G1 ACTIAS21871G5
1-Acetyl-4-bromo-1H-indazole
1-Acetyl-4-bromo-1H-indazole
Formula: C₉H₇BrN₂O
MDL Number: MFCD12198857
CAS Number: 885698-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR