Diethylzinc 1 mol/l in heptane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

406023-100ML 406023-800ML
SIAL406023-100MLEA 115 GBP
SIAL406023-100ML SIAL406023-800ML
Diethylzinc 1 mol/l in heptane, Sigma-Aldrich®
Diethylzinc
Formula: C₄H₁₀Zn
MW: 123.51 g/mol
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR