2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1255.1 CFF1255.5
CHMPCFF1255.1EA 0 GBP
CHMPCFF1255.1 CHMPCFF1255.5
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
MDL Number: MFCD00269226
CAS Number: 123533-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR