Ethyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1096.1 CFF1096.5 CFF1096.25
CHMPCFF1096.1EA 0 GBP
CHMPCFF1096.1 CHMPCFF1096.5 CHMPCFF1096.25
Ethyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₆H₁₄N₂O₂
MDL Number: MFCD00139520
CAS Number: 119448-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR