2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF965.1 CFF965.5
CHMPCFF965.1EA 0 GBP
CHMPCFF965.1 CHMPCFF965.5
2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride
2-(Chloromethyl)imidazo[1,2-a]pyridine hydrochloride
Formula: C₈H₈Cl₂N₂
MW: 203.07 g/mol
MDL Number: MFCD03094615
CAS Number: 112230-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR