Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate
AS2234G1
AS2234G5
ACTIAS2234G1EA
115
GBP
ACTIAS2234G1
ACTIAS2234G5
Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₀H₉IN₂O₂ MW: 316.1000 g/mol |
MDL Number:
MFCD02186151 CAS Number: 214958-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...