Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

PY-7155-1G PY-7155-5G
COBBPY-7155-1GEA 0 GBP
COBBPY-7155-1G COBBPY-7155-5G
Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉ClN₂O₂
MW: 224.6437 g/mol
MDL Number: MFCD02186220
CAS Number: 67625-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR