Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

AS12096G1 AS12096G5
ACTIAS12096G1EA 50 GBP
ACTIAS12096G1 ACTIAS12096G5
Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉ClN₂O₂
MW: 224.6437 g/mol
MDL Number: MFCD02186220
CAS Number: 67625-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR