Imidazo[1,2-a]pyridine-3-carbaldehyde
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6659-5G
HC-6659-1G
COBBHC-6659-5GEA
0
GBP
COBBHC-6659-5G
COBBHC-6659-1G
Imidazo[1,2-a]pyridine-3-carbaldehyde
Imidazo[1,2-a]pyridine-3-carbaldehyde
Formula:
C₈H₆N₂O MW: 146.1500 g/mol |
MDL Number:
MFCD07778354 CAS Number: 6188-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...