Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

HC-6501-5G HC-6501-1G
COBBHC-6501-5GEA 0 GBP
COBBHC-6501-5G COBBHC-6501-1G
Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate
Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate
Formula: C₉H₈ClN₃O₂
MW: 225.6317 g/mol
MDL Number: MFCD03086230
CAS Number: 64067-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR