2-(Ethylsulphonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8394-1G BB-8394-250M
COBBBB-8394-1GEA 0 GBP
COBBBB-8394-1G COBBBB-8394-250M
2-(Ethylsulphonyl)phenylboronic acid
2-(Ethylsulphonyl)phenylboronic acid
Formula: C₈H₁₁BO₄S
MDL Number: MFCD03095198
CAS Number: 1042443-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR