Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate

PY-7098-5G PY-7098-1G
COBBPY-7098-5GEA 0 GBP
COBBPY-7098-5G COBBPY-7098-1G
Ethyl-6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉BrN₂O₂
MW: 269.10 g/mol
MDL Number: MFCD06659034
CAS Number: 67625-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR